missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(3,5-Difluorophenyl)propionic acid, 97%

Manufacturer:  Alfa Aesar™ H5400406

 View more versions of this product

Catalog No. AAH5400406



Description

Description

Applications
3-(3,5-Difluorophenyl)propionic acid is used as intermediate in pharmaceuticals.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications

Specifications

3-(3,5-Difluorophenyl)propionic acid
5g
C9H8F2O2
3,5-difluoro-benzenepropanoic acid
C1=C(C=C(C=C1F)F)CCC(=O)O
186.158
186.16
60°C
84315-24-2
MFCD04116054
SAAKANGUQMVTHQ-UHFFFAOYSA-N
3-(3,5-difluorophenyl)propanoic acid
3669498
97%

Chemical Identifiers

84315-24-2
186.158
SAAKANGUQMVTHQ-UHFFFAOYSA-N
3669498
C1=C(C=C(C=C1F)F)CCC(=O)O
C9H8F2O2
MFCD04116054
3,5-difluoro-benzenepropanoic acid
3-(3,5-difluorophenyl)propanoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H311-H318-H315-H335
Toxic in contact with skin.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c

H302+H312-H315-H318-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents