Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(3,5-Difluorophenyl)-1H-pyrazole-4-carboxaldehyde, 98%

$45.51 - $45.51

Chemical Identifiers

Chemical Name or Material 3-(3,5-Difluorophenyl)-1H-pyrazole-4-carboxaldehyde
CAS 875664-59-8
Molecular Formula C10H6F2N2O
Formula Weight 208.16
MDL Number MFCD06797905
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH26132MD

  SDS
alfa aesar™
H26132MD
250mg Each for $45.51
Description & Specifications

Specifications

Formula Weight 208.16
Quantity 250mg
Molecular Formula C10H6F2N2O
CAS 875664-59-8
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 3-(3,5-Difluorophenyl)-1H-pyrazole-4-carboxaldehyde
Assay 98%
MDL Number MFCD06797905
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 178° to 181°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.