missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(3,5-Dichlorophenyl)propionic acid, 97%

$116.20 - $516.51

Chemical Identifiers

CAS 95333-95-2
Molecular Formula C9H8Cl2O2
Molecular Weight (g/mol) 219.061
MDL Number MFCD04037332
InChI Key GQICJNXGCGRFBA-UHFFFAOYSA-N
Synonym 3,5-dichloro-benzenepropanoic acid
PubChem CID 819305
IUPAC Name 3-(3,5-dichlorophenyl)propanoic acid
SMILES C1=C(C=C(C=C1Cl)Cl)CCC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5400906 View Documents Alfa Aesar™
H5400906
5g Each for $116.20
 
AAH5400914 View Documents Alfa Aesar™
H5400914
25g Each for $516.51
 
Description

Description

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications

Chemical Identifiers

95333-95-2
219.061
GQICJNXGCGRFBA-UHFFFAOYSA-N
819305
C1=C(C=C(C=C1Cl)Cl)CCC(=O)O
C9H8Cl2O2
MFCD04037332
3,5-dichloro-benzenepropanoic acid
3-(3,5-dichlorophenyl)propanoic acid

Specifications

56°C to 60°C
95333-95-2
MFCD04037332
Slightly soluble in water.
C1=C(C=C(C=C1Cl)Cl)CCC(=O)O
219.061
219.07
3-(3,5-Dichlorophenyl)propionic acid
5g
C9H8Cl2O2
3,5-dichloro-benzenepropanoic acid
GQICJNXGCGRFBA-UHFFFAOYSA-N
3-(3,5-dichlorophenyl)propanoic acid
819305
97%
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures