missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(3,5-Dichlorophenyl)propionic acid, 97%

Manufacturer:  Alfa Aesar™ H5400914

 View more versions of this product

Catalog No. AAH5400914



Description

Description

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications

Specifications

3-(3,5-Dichlorophenyl)propionic acid
25g
C9H8Cl2O2
3,5-dichloro-benzenepropanoic acid, 3-3',5'-dichlorophenyl propanoic acid, 3-3,5-di-chlorophenyl propionic acid, 3-3,5-dichloro-phenyl-propionic acid, 3-3,5-dichloro-phenyl-propionicacid, 3-3,5-dichlorophenyl propanoic acid, 3-3,5-dichlorophenyl propanoicacid, 3-3,5-dichlorophenyl propionic acid, acmc-20anl7, benzenepropanoic acid, 3,5-dichloro
GQICJNXGCGRFBA-UHFFFAOYSA-N
3-(3,5-dichlorophenyl)propanoic acid
819305
97%
56°C to 60°C
95333-95-2
MFCD04037332
Slightly soluble in water.
C1=C(C=C(C=C1Cl)Cl)CCC(=O)O
219.061
219.07

Chemical Identifiers

95333-95-2
219.061
GQICJNXGCGRFBA-UHFFFAOYSA-N
819305
C1=C(C=C(C=C1Cl)Cl)CCC(=O)O
C9H8Cl2O2
MFCD04037332
3,5-dichloro-benzenepropanoic acid, 3-3',5'-dichlorophenyl propanoic acid, 3-3,5-di-chlorophenyl propionic acid, 3-3,5-dichloro-phenyl-propionic acid, 3-3,5-dichloro-phenyl-propionicacid, 3-3,5-dichlorophenyl propanoic acid, 3-3,5-dichlorophenyl propanoicacid, 3-3,5-dichlorophenyl propionic acid, acmc-20anl7, benzenepropanoic acid, 3,5-dichloro
3-(3,5-dichlorophenyl)propanoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents