missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,3′,5,5′-Tetramethylbiphenyl-4,4′-diol, TCI America™

Manufacturer:  TCI America T314525G

 View more versions of this product

Catalog No. T314525G



Specifications

Specifications

3,3′,5,5′-Tetramethylbiphenyl-4,4′-diol
225°C
2417-04-1
MFCD00094737
YGYPMFPGZQPETF-UHFFFAOYSA-N
4-(4-hydroxy-3,5-dimethylphenyl)-2,6-dimethylphenol
75490
Crystal-Powder at 20°C
Yellow
25g
C16H18O2
4,4′-Dihydroxy-3,3′,5,5′-tetramethylbiphenyl
CC1=CC(=CC(=C1O)C)C2=CC(=C(C(=C2)C)O)C
242.32
242.32
≥98.0% (GC)

Chemical Identifiers

2417-04-1
242.32
YGYPMFPGZQPETF-UHFFFAOYSA-N
75490
CC1=CC(=CC(=C1O)C)C2=CC(=C(C(=C2)C)O)C
C16H18O2
MFCD00094737
4,4′-Dihydroxy-3,3′,5,5′-tetramethylbiphenyl
4-(4-hydroxy-3,5-dimethylphenyl)-2,6-dimethylphenol
Safety and Handling

Safety and Handling

EINECSNumber : (4)-1675

TSCA : Yes

Documents