missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,3',5,5'-Tetramethylbenzidine soln., Ready-to-Use, standard sensitivity

Manufacturer:  Alfa Aesar™ J63202AU

 View more versions of this product

Catalog No. AAJ63202AU



Description

Description

  • Excellent colorimetric substrate for detection of horseradish peroxidase labelled probes and is used with peroxidase and peroxidase coupled systems, particularly in ELISA techniques, blue result
Specifications

Specifications

3,3',5,5'-Tetramethylbenzidine soln.
1000mL
54827-17-7
MFCD00007748
3,3',5,5'-tetramethyl benzidine, 3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine, 3,3',5,5'-tetramethylbenzidine, 3,5,3',5'-tetramethylbenzidine, bm blue, ccris 4727, tmb, tmb plus, tmb substrate, tmb, insoluble
CC1=CC(=CC(=C1N)C)C2=CC(=C(C(=C2)C)N)C
240.35
240.35
Yellow
Ready-to-Use, standard sensitivity
C16H20N2
2808541
UAIUNKRWKOVEES-UHFFFAOYSA-N
4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline
41206
Liquid

Chemical Identifiers

54827-17-7
240.35
UAIUNKRWKOVEES-UHFFFAOYSA-N
41206
CC1=CC(=CC(=C1N)C)C2=CC(=C(C(=C2)C)N)C
C16H20N2
MFCD00007748
3,3',5,5'-tetramethyl benzidine, 3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine, 3,3',5,5'-tetramethylbenzidine, 3,5,3',5'-tetramethylbenzidine, bm blue, ccris 4727, tmb, tmb plus, tmb substrate, tmb, insoluble
4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P307+P311-P311-P312-P330-P363-P370+P378q-P501c

H226-H301+H331-H312-H336-H370

DOTInformation : Hazard Class: 3; Packaging Group: II

EINECSNumber : 259-364-6

TSCA : Yes

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

Documents