Learn More
3,3',5,5'-Tetramethylbenzidine soln., Ready-to-Use, precipitating, high sensitivity, Thermo Scientific Chemicals
$381.63 - $381.63
Chemical Identifiers
CAS | 54827-17-7 |
---|---|
Molecular Formula | C16H20N2 |
Molecular Weight (g/mol) | 240.35 |
MDL Number | MFCD00007748 |
InChI Key | UAIUNKRWKOVEES-UHFFFAOYSA-N |
Synonym | 3,3',5,5'-tetramethylbenzidine, 3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine, 3,5,3',5'-tetramethylbenzidine, tmb, 3,3',5,5'-tetramethyl benzidine, tmb substrate, bm blue, ccris 4727, tmb plus, tmb, insoluble |
PubChem CID | 41206 |
IUPAC Name | 4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline |
SMILES | CC1=CC(=CC(=C1N)C)C2=CC(=C(C(=C2)C)N)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ62143AP
|
Thermo Scientific Chemicals
J62143AP |
500 mL |
Each for $381.63
|
|
|||||
Description
Excellent colorimetric substrate for detection of horseradish peroxidase labelled probes and is used with peroxidase and peroxidase coupled systems, particularly in ELISA techniques, blue result
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Excellent colorimetric substrate for detection of horseradish peroxidase labelled probes and is used with peroxidase and peroxidase-coupled systems, particularly in ELISA techniques, blue result
Chemical Identifiers
54827-17-7 | |
240.35 | |
UAIUNKRWKOVEES-UHFFFAOYSA-N | |
41206 | |
CC1=CC(=CC(=C1N)C)C2=CC(=C(C(=C2)C)N)C |
C16H20N2 | |
MFCD00007748 | |
3,3',5,5'-tetramethylbenzidine, 3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine, 3,5,3',5'-tetramethylbenzidine, tmb, 3,3',5,5'-tetramethyl benzidine, tmb substrate, bm blue, ccris 4727, tmb plus, tmb, insoluble | |
4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline |
Specifications
54827-17-7 | |
Alcohol-like | |
MFCD00007748 | |
UN2810 | |
3,3',5,5'-tetramethylbenzidine, 3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine, 3,5,3',5'-tetramethylbenzidine, tmb, 3,3',5,5'-tetramethyl benzidine, tmb substrate, bm blue, ccris 4727, tmb plus, tmb, insoluble | |
CC1=CC(=CC(=C1N)C)C2=CC(=C(C(=C2)C)N)C | |
240.35 | |
240.35 | |
3,3',5,5'-Tetramethylbenzidine soln., Ready-to-Use, precipitating, high sensitivity |
Colorless to Yellow | |
C16H20N2 | |
500 mL | |
2808541 | |
UAIUNKRWKOVEES-UHFFFAOYSA-N | |
4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline | |
41206 | |
Liquid |
Safety and Handling
GHS H Statement
H370-H312-H332
Causes damage to organs.
Harmful in contact with skin.
Harmful if inhaled.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P307+P311-P311-P312-P330-P363-P370+P378q-P501c
H226-H302+H312-H331-H336-H370
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber : 259-364-6
TSCA : Yes
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only