Learn More
3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate, 98+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 229990050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate | |
98% min. (on dry substance) (Argentometry) | |
2HCl·xH2O | |
3,3',5,5'-tetramethylbenzidine dihydrochloride dihydrate, 3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine dihydrochloride dihydrate, 3,3',5,5'-tetramethylbenzidine dihydrochloride hydrate, pubchem9044, tetramethylbenzidine dihydrate dihydrochloride | |
CJRXJBONCVPXPJ-UHFFFAOYSA-N | |
4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline;dihydrate;dihydrochloride | |
19836601 | |
98+% |
207738-08-7 | |
2HCl·xH2O | |
5g | |
Solubility in water: moderately soluble. Other solubilities: soluble in methanol | |
CC1=CC(=CC(=C1N)C)C2=CC(=C(C(=C2)C)N)C.O.O.Cl.Cl | |
313.26 | |
313.26 |
Chemical Identifiers
207738-08-7 | |
313.26 | |
3,3',5,5'-tetramethylbenzidine dihydrochloride dihydrate, 3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine dihydrochloride dihydrate, 3,3',5,5'-tetramethylbenzidine dihydrochloride hydrate, pubchem9044, tetramethylbenzidine dihydrate dihydrochloride | |
4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline;dihydrate;dihydrochloride |
2HCl·xH2O | |
CJRXJBONCVPXPJ-UHFFFAOYSA-N | |
19836601 | |
CC1=CC(=CC(=C1N)C)C2=CC(=C(C(=C2)C)N)C.O.O.Cl.Cl |
Safety and Handling
GHS H Statement May cause damage to organs. Harmful in contact with skin. Suspected of causing genetic defects. Harmful if inhaled. Harmful if swallowed.
GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Use personal protective equipment as required. IF ON SKIN: Wash with plenty of soap and water. Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Warning
RUO â Research Use Only