Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,3',5,5'-Tetramethylbenzidine aq. soln.

Chemical Identifiers

Chemical Name or Material 3,3',5,5'-Tetramethylbenzidine
CAS 54827-17-7
Molecular Formula C16H20N2
Formula Weight 240.35
MDL Number MFCD00007748
Beilstein 2808541
Synonym TMB soluble reagent, aq. soln.
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ60808EQE

  SDS
alfa aesar™
J60808EQE
125mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 240.35
Chemical Name or Material 3,3',5,5'-Tetramethylbenzidine
TSCA TSCA
Physical Form Liquid
Molecular Formula C16H20N2
MDL Number MFCD00007748
CAS 54827-17-7
Beilstein 2808541
Synonym TMB soluble reagent, aq. soln.
Color Pale Yellow
Quantity 125mL
Recommended Storage 2° to 8°C

Applications
3,3’,5,5’-Tetramethylbenzidine or TMB is a chromogenic substrate used in staining procedures in immunohistochemistry as well as being a visualising reagent used in enzyme-linked immunosorbent assays. 3,3′,5,5′-Tetramethylbenzidine (TMB) is a noncarcinogenic substitute (Ames test negative) for benzidine suitable as peroxidase substrate for use in ELISA procedures. The substrate produces a soluble end product that is pale blue in color and can be read spectrophotometrically at 370 or 620-650 nm. The TMB reaction may be stopped with 2 M H2SO4 (resulting in a yellow color), and read at 450 nm. A sensitive and specific reagent for the detection of blood assay of hemoglobin assay of peroxidases.

Solubility
Slightly soluble. <0.1 g/100 mL at 20°C.

Notes
Contains TMB in an aqueous buffer, pH 3.6 ± 0.2, with 0.03% hydrogen peroxide. The TMB product is soluble after conversion

Safety and Handling

2° to 8°C