missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,3',5,5'-Tetramethylbenzidine, 99+%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC229280010
Specifications
3,3′,5,5′-Tetramethylbenzidine | |
0.5% max. | |
0.2% max. | |
167.0°C to 170.0°C | |
54827-17-7 | |
C16H20N2 | |
3,3',5,5'-tetramethyl benzidine, 3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine, 3,3',5,5'-tetramethylbenzidine, 3,5,3',5'-tetramethylbenzidine, bm blue, ccris 4727, tmb, tmb plus, tmb substrate, tmb, insoluble | |
UAIUNKRWKOVEES-UHFFFAOYSA-N | |
4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline | |
41206 | |
Crystalline Powder |
Authentic | |
Glass bottle | |
Cream | |
1g | |
99% min. (HPLC) | |
(H2N(CH3)2C6H2-)2 | |
Solubility in water: insoluble. Other solubilities: readily soluble in methanol, soluble in solvents | |
CC1=CC(=CC(=C1N)C)C2=CC(=C(C(=C2)C)N)C | |
240.35 | |
240.35 | |
99+% |
Chemical Identifiers
54827-17-7 | |
240.35 | |
3,3',5,5'-tetramethyl benzidine, 3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine, 3,3',5,5'-tetramethylbenzidine, 3,5,3',5'-tetramethylbenzidine, bm blue, ccris 4727, tmb, tmb plus, tmb substrate, tmb, insoluble | |
4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline |
C16H20N2 | |
UAIUNKRWKOVEES-UHFFFAOYSA-N | |
41206 | |
CC1=CC(=CC(=C1N)C)C2=CC(=C(C(=C2)C)N)C |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plen
GHS Signal Word: Warning
EINECSNumber : 259-364-6