Learn More
3,3',5,5'-Tetramethylbenzidine, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 229280050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3,3′,5,5′-Tetramethylbenzidine | |
167.0°C to 170.0°C | |
0.5% max. | |
99% min. (HPLC) | |
C16H20N2 | |
5 g | |
Solubility in water: insoluble. Other solubilities: readily soluble in methanol, soluble in solvents | |
CC1=CC(=CC(=C1N)C)C2=CC(=C(C(=C2)C)N)C | |
240.35 | |
240.35 | |
Crystalline Powder |
54827-17-7 | |
Cream | |
Authentic | |
Glass bottle | |
(H2N(CH3)2C6H2-)2 | |
3,3',5,5'-tetramethylbenzidine, 3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine, 3,5,3',5'-tetramethylbenzidine, tmb, 3,3',5,5'-tetramethyl benzidine, tmb substrate, bm blue, ccris 4727, tmb plus, tmb, insoluble | |
UAIUNKRWKOVEES-UHFFFAOYSA-N | |
4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline | |
41206 | |
99+% |
Chemical Identifiers
54827-17-7 | |
240.35 | |
3,3',5,5'-tetramethylbenzidine, 3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine, 3,5,3',5'-tetramethylbenzidine, tmb, 3,3',5,5'-tetramethyl benzidine, tmb substrate, bm blue, ccris 4727, tmb plus, tmb, insoluble | |
4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline |
C16H20N2 | |
UAIUNKRWKOVEES-UHFFFAOYSA-N | |
41206 | |
CC1=CC(=CC(=C1N)C)C2=CC(=C(C(=C2)C)N)C |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plen
GHS Signal Word: Warning
EINECSNumber : 259-364-6
RUO – Research Use Only