missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,3',5,5'-Tetrakis(trifluoromethyl)benzophenone, 98%

Manufacturer:  Alfa Aesar™ A1116406

 View more versions of this product

Catalog No. AAA1116406



Description

Description

Applications
3,3′,5,5′-Tetrakis(trifluoromethyl)benzophenone, is used as reactive dye intermediates.

Solubility
Sparingly soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications

Specifications

3,3',5,5'-Tetrakis (trifluoromethyl)benzophenone
98%
MFCD00042474
C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C(=O)C2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F
454.215
454.22
175136-66-0
C17H6F12O
GATWMPGNBWPCIY-UHFFFAOYSA-N
bis[3,5-bis(trifluoromethyl)phenyl]methanone
519439

Chemical Identifiers

175136-66-0
454.215
GATWMPGNBWPCIY-UHFFFAOYSA-N
bis[3,5-bis(trifluoromethyl)phenyl]methanone
C17H6F12O
MFCD00042474
519439
C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C(=O)C2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents