Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,3',5,5'-Tetrakis(trifluoromethyl)benzhydrol, 97%

$235.05 - $973.36

Chemical Identifiers

Chemical Name or Material 3,3',5,5'-Tetrakis(trifluoromethyl)benzhydrol
CAS 87901-76-6
Molecular Formula C17H8F12O
Formula Weight 456.23
MDL Number MFCD00042475
Synonym Bis[3,5-bis(trifluoromethyl)phenyl] carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2055306

  SDS
alfa aesar™
B2055306
5g Each for $235.05

AAB2055314

  SDS
alfa aesar™
B2055314
25g Each for $973.36
Description & Specifications

Specifications

Formula Weight 456.23
Molecular Formula C17H8F12O
Quantity 5g
Synonym Bis[3,5-bis(trifluoromethyl)phenyl] carbinol
CAS 87901-76-6
Melting Point 147° to 149°C
Chemical Name or Material 3,3',5,5'-Tetrakis(trifluoromethyl)benzhydrol
Assay 97%
MDL Number MFCD00042475