Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,3',5,5'-Tetraiodophenolsulfonephthalein

$219.99 - $219.99

Chemical Identifiers

Chemical Name or Material 3,3',5,5'-Tetraiodophenolsulfonephthalein
CAS 4430-24-4
Molecular Formula C19H10I4O5S
Formula Weight 857.97
MDL Number MFCD00010405
Beilstein 366159
Synonym Iodophenol Blue
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2423614

  SDS
alfa aesar™
B2423614
25g Each for $219.99
Description & Specifications

Specifications

Formula Weight 857.97
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C19H10I4O5S
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
CAS 4430-24-4
Melting Point ≈229°C (dec.)
Quantity 25g
Chemical Name or Material 3,3',5,5'-Tetraiodophenolsulfonephthalein
MDL Number MFCD00010405
Beilstein 366159
Synonym Iodophenol Blue
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.