Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,3',5,5'-Tetrabromobisphenol A, 97%

Chemical Identifiers

Chemical Name or Material 3,3',5,5'-Tetrabromobisphenol A
CAS 79-94-7
Molecular Formula C15H12Br4O2
Formula Weight 543.87
MDL Number MFCD00013962
UN Number UN3077
Synonym 2,2-Bis(3,5-dibromo-4-hydroxyphenyl)propane; 4,4'-Isopropylidenebis(2,6-dibromophenol)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA173950E

  SDS
alfa aesar™
A173950E
2500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1739522

  SDS
alfa aesar™
A1739522
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1739536

  SDS
alfa aesar™
A1739536
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 543.87
Chemical Name or Material 3,3',5,5'-Tetrabromobisphenol A
TSCA TSCA
Assay 97%
Molecular Formula C15H12Br4O2
MDL Number MFCD00013962
CAS 79-94-7
UN Number UN3077
Synonym 2,2-Bis(3,5-dibromo-4-hydroxyphenyl)propane; 4,4'-Isopropylidenebis(2,6-dibromophenol)
Melting Point 179° to 182°C
Odor Odorless
Quantity 2500g
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
RTECS Number SM0894500