missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,3',5,5'-Tetrabromo-2,2'-bithiophene 98.0+%, TCI America™
$69.68 - $622.13
Chemical Identifiers
CAS | 125143-53-5 |
---|---|
Molecular Formula | C8H2Br4S2 |
Molecular Weight (g/mol) | 481.84 |
InChI Key | MOMHMPZSZNZLAK-UHFFFAOYSA-N |
PubChem CID | 2740586 |
IUPAC Name | 3,5-dibromo-2-(3,5-dibromothiophen-2-yl)thiophene |
SMILES | C1=C(SC(=C1Br)C2=C(C=C(S2)Br)Br)Br |
Chemical Identifiers
125143-53-5 | |
481.84 | |
2740586 | |
C1=C(SC(=C1Br)C2=C(C=C(S2)Br)Br)Br |
C8H2Br4S2 | |
MOMHMPZSZNZLAK-UHFFFAOYSA-N | |
3,5-dibromo-2-(3,5-dibromothiophen-2-yl)thiophene |
Specifications
125143-53-5 | |
C8H2Br4S2 | |
MOMHMPZSZNZLAK-UHFFFAOYSA-N | |
3,5-dibromo-2-(3,5-dibromothiophen-2-yl)thiophene | |
2740586 | |
≥98.0% (GC) | |
3,3′,5,5′-Tetrabromo-2,2′-bithiophene |
140°C | |
1 g | |
C1=C(SC(=C1Br)C2=C(C=C(S2)Br)Br)Br | |
481.84 | |
481.84 | |
Crystalline Powder |
Safety and Handling
TSCA : No