Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(3,4-Methylenedioxyphenyl)propionic acid, 99%

$1,455.53 - $1,455.53

Chemical Identifiers

Chemical Name or Material 3-(3,4-Methylenedioxyphenyl)propionic acid
CAS 2815-95-4
Molecular Formula C10H10O4
Formula Weight 194.19
MDL Number MFCD00016909
Beilstein 181735
Synonym 3-(1,3-Benzodioxyl-5-yl)propionic acid; 3,4-(Methylenedioxy)hydrocinnamic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1476322

  SDS
alfa aesar™
A1476322
100g Each for $1,455.53
Description & Specifications

Specifications

Formula Weight 194.19
Assay 99%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C10H10O4
Beilstein 181735
Quantity 100g
Chemical Name or Material 3-(3,4-Methylenedioxyphenyl)propionic acid
Health Hazard 1 Warning
Melting Point 86° to 88°C
Synonym 3-(1,3-Benzodioxyl-5-yl)propionic acid; 3,4-(Methylenedioxy)hydrocinnamic acid
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00016909
CAS 2815-95-4
RTECS Number MW5600000

Applications
3-(3,4-Methylenedioxyphenyl)propionic acid exhibits anti-inflammatory and analgesic activity. Further, it is used in the preparation of 3-benzo[1,3]dioxol-5-yl-propionyl chloride by reaction with thionyl chloride.

Solubility
Soluble in chloroform and ethyl acetate.

Notes
Incompatible with strong oxidizing agents, strong reducing agents, strong bases and amines.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.