Please login to your online account to display your discounted pricing

3-(3,4-Dimethoxyphenyl)propionic acid, 99%, ACROS Organics™

$314.20 - $314.20

Chemical Identifiers

Chemical Name or Material 3-(3, 4-Dimethoxyphenyl)propionic acid
Name Note 99%
CAS 2107-70-2
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C11H14O4
Linear Formula (CH3O)2C6H3CH2CH2CO2H
Formula Weight 210.23
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC171910500

  SDS
thermo fisher scientific
171910500
Glass bottle 50g Each for $314.20
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 210.23
Quantity 50g
Color Slightly beige
Beilstein 10, 424
EINECS Number 218-288-3
TSCA TSCA
Melting Point 97° to 100°C
CAS Max % 100.0
Chemical Name or Material 3-(3, 4-Dimethoxyphenyl)propionic acid
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 99%
Assay Percent Range 99%
Molecular Formula C11H14O4
Linear Formula (CH3O)2C6H3CH2CH2CO2H
MDL Number MFCD00002774
CAS 2107-70-2
Synonym 3, 4-Dimethoxyhydrocinnamic acid
CAS Min % 98.5