missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(3,4-Dihydroxyphenyl)-L-alanine, 99%, ACROS Organics™

Manufacturer:  Acros Organics AC167535000

 View more versions of this product

Catalog No. AC167535000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3-(3, 4-Dihydroxyphenyl)-L-alanine
99%
C9H11NO4
MFCD00002598
1% max.
Glass bottle
WTDRDQBEARUVNC-LURJTMIESA-N
(2S)-2-amino-3-(3,4-dihydroxyphenyl)propanoic acid
6047
197.19
≥98.5%
Cream to White
59-92-7
Authentic
(HO)2C6H3CH2CH(NH2)CO2H
14, IV, 2492
15, 5516
levodopa, l-dopa, dopar, 3-hydroxy-l-tyrosine, 3,4-dihydroxy-l-phenylalanine, larodopa, bendopa, levopa, dihydroxy-l-phenylalanine, cidandopa
C1=CC(=C(C=C1CC(C(=O)O)N)O)O
197.19
CHEBI:15765
Crystalline Powder
0.1% max.
500g

Chemical Identifiers

59-92-7
197.19
WTDRDQBEARUVNC-LURJTMIESA-N
6047
(2S)-2-amino-3-(3,4-dihydroxyphenyl)propanoic acid
C9H11NO4
MFCD00002598
levodopa, l-dopa, dopar, 3-hydroxy-l-tyrosine, 3,4-dihydroxy-l-phenylalanine, larodopa, bendopa, levopa, dihydroxy-l-phenylalanine, cidandopa
CHEBI:15765
C1=CC(=C(C=C1CC(C(=O)O)N)O)O
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS Documents