Learn More
3-(3,4-Dichlorophenyl)-1,1-dimethylurea, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0298618
Description
A phenyl urea pre-emergent agrochemical. Also an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyestuff
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsA phenyl urea pre-emergent agrochemical. Also an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyestuff
Solubility
Soluble in water (0.049g/L), chloroform and methanol. Very slightly soluble in many organic solvents.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
3-(3,4-Dichlorophenyl)-1,1-dimethylurea | |
157°C to 160°C | |
180°C to 190°C (decomposition) | |
C9H10Cl2N2O | |
50 g | |
2215168 | |
diuron, 3-3,4-dichlorophenyl-1,1-dimethylurea, dcmu, dynex, duran, dichlorfenidim, herbatox, vonduron, dailon, karmex | |
XMTQQYYKAHVGBJ-UHFFFAOYSA-N | |
3-(3,4-dichlorophenyl)-1,1-dimethylurea | |
3120 | |
233.1 |
330-54-1 | |
1.48 | |
Odorless | |
MFCD00018136 | |
UN3077 | |
14,3382 | |
Soluble in water (0.049g/L),chloroform and methanol. Very slightly soluble in many organic solvents. | |
CN(C)C(=O)NC1=CC(=C(C=C1)Cl)Cl | |
233.092 | |
CHEBI:116509 | |
97% |
Chemical Identifiers
330-54-1 | |
233.092 | |
XMTQQYYKAHVGBJ-UHFFFAOYSA-N | |
3120 | |
3-(3,4-dichlorophenyl)-1,1-dimethylurea |
C9H10Cl2N2O | |
MFCD00018136 | |
diuron, 3-3,4-dichlorophenyl-1,1-dimethylurea, dcmu, dynex, duran, dichlorfenidim, herbatox, vonduron, dailon, karmex | |
CHEBI:116509 | |
CN(C)C(=O)NC1=CC(=C(C=C1)Cl)Cl |
Safety and Handling
GHS H Statement
H351-H373-H302
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
P201-P202-P260-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
H302-H351-H373
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 206-354-4
RTECSNumber : YS8925000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only