Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,3',4,4'-Tetramethylbiphenyl, 98%

$75.88 - $75.88

Chemical Identifiers

Chemical Name or Material 3,3',4,4'-Tetramethylbiphenyl
CAS 4920-95-0
Molecular Formula C16H18
Formula Weight 210.32
MDL Number MFCD00130219
Beilstein 1935370
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1333803

 
alfa aesar™
L1333803
1g Each for $75.88
Description & Specifications

Specifications

Formula Weight 210.32
Chemical Name or Material 3,3',4,4'-Tetramethylbiphenyl
Molecular Formula C16H18
Assay 98%
MDL Number MFCD00130219
CAS 4920-95-0
RTECS Number DV8730000
TSCA TSCA
Beilstein 1935370
Melting Point 74° to 77°C
Quantity 1g