Please login to your online account to display your discounted pricing

3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluoro-1-octene, 99%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  309010250

 View more versions of this product

Catalog No. AC309010250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Density 1.5200g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.2920 to 1.2950 (20°C, 589nm)
Color Clear colorless
Quantity 25g
Chemical Name or Material 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluoro-1-octene
CAS 25291-17-2
CAS Min % 98.5
CAS Max % 100.0
Physical Form Clear colorless liquid
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C8H3F13
Linear Formula CF3(CF2)4CF2CH=CH2
Formula Weight 346.09g/mol
MDL Number MFCD00039249
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluoro-1-octene
CAS 25291-17-2
CAS Min % 98.5
CAS Max % 100.0
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C8H3F13
Linear Formula CF3(CF2)4CF2CH=CH2
Formula Weight 346.09g/mol
MDL Number MFCD00039249
Accessories

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Highly flammable liquid and vapor.

SDS & Safety Documents