Please login to your online account to display your discounted pricing

3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluoro-1-octene, 99%, ACROS Organics™

$38.09 - $38.09

Chemical Identifiers

Chemical Name or Material 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluoro-1-octene
CAS 25291-17-2
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H3F13
Linear Formula CF3(CF2)4CF2CH=CH2
Formula Weight 346.09
MDL Number MFCD00039249
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC309010250

  SDS
thermo fisher scientific
AC309010250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC309010050

  SDS
thermo fisher scientific
309010050
Glass bottle 5g Each for $38.09
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Liquid
Formula Weight 346.09
Quantity 25g
Color Clear colorless
Density 1.5200g/mL
CAS Max % 100.0
Chemical Name or Material 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluoro-1-octene
Molecular Formula C8H3F13
Assay Percent Range 99%
CAS 25291-17-2
CAS Min % 98.5
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Linear Formula CF3(CF2)4CF2CH=CH2
Infrared Spectrum Authentic
MDL Number MFCD00039249
Health Hazard 1 GHS Signal Word: Danger
Packaging Glass bottle
Refractive Index 1.2920 to 1.2950 (20°C, 589nm)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.

Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Highly flammable liquid and vapor.