Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,3,4,4,5,5,5-Heptafluoro-2-pentanone, 97%

Chemical Identifiers

Chemical Name or Material 3,3,4,4,5,5,5-Heptafluoro-2-pentanone
CAS 355-17-9
Molecular Formula C5H3F7O
Formula Weight 212.07
MDL Number MFCD00042079
UN Number UN1224
Synonym Heptafluoropropyl methyl ketone; Methyl heptafluoropropyl ketone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1680403

 
alfa aesar™
L1680403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1680406

 
alfa aesar™
L1680406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 212.07
Chemical Name or Material 3,3,4,4,5,5,5-Heptafluoro-2-pentanone
Assay 97%
Molecular Formula C5H3F7O
MDL Number MFCD00042079
CAS 355-17-9
Synonym Heptafluoropropyl methyl ketone; Methyl heptafluoropropyl ketone
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 63° to 64°C
Quantity 1g
UN Number UN1224
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: KETONES, LIQUID, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.