missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,3,4,4,5,5,5-Heptafluoro-2-pentanol, 96%

$36.39 - $123.51

Chemical Identifiers

CAS 375-14-4
Molecular Formula C5H5F7O
Molecular Weight (g/mol) 214.083
MDL Number MFCD00039619
InChI Key RBPHBIMHZSTIDT-UHFFFAOYSA-N
Synonym 3,3,4,4,5,5,5-heptafluoro-2-pentanol, acmc-20anhf, 3,3,4,4,5,5,5-heptafluoropentanol-2, 2-pentanol,3,3,4,4,5,5,5-heptafluoro, 2-pentanol, 3,3,4,4,5,5,5-heptafluoro, 3,3,4,4,5,5,5-heptafluoro-2-pentanol, 3,3,4,4,5,5,5-heptakis fluoranyl pentan-2-ol
PubChem CID 78992
IUPAC Name 3,3,4,4,5,5,5-heptafluoropentan-2-ol
SMILES CC(C(C(C(F)(F)F)(F)F)(F)F)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1678203 View Documents Alfa Aesar™
L1678203
1g Each for $36.39
 
AAL1678206 View Documents Alfa Aesar™
L1678206
5g Each for $123.51
 
Specifications

Chemical Identifiers

375-14-4
214.083
RBPHBIMHZSTIDT-UHFFFAOYSA-N
78992
CC(C(C(C(F)(F)F)(F)F)(F)F)O
C5H5F7O
MFCD00039619
3,3,4,4,5,5,5-heptafluoro-2-pentanol, acmc-20anhf, 3,3,4,4,5,5,5-heptafluoropentanol-2, 2-pentanol,3,3,4,4,5,5,5-heptafluoro, 2-pentanol, 3,3,4,4,5,5,5-heptafluoro, 3,3,4,4,5,5,5-heptafluoro-2-pentanol, 3,3,4,4,5,5,5-heptakis fluoranyl pentan-2-ol
3,3,4,4,5,5,5-heptafluoropentan-2-ol

Specifications

101°C to 102°C
375-14-4
MFCD00039619
3,3,4,4,5,5,5-heptafluoro-2-pentanol, acmc-20anhf, 3,3,4,4,5,5,5-heptafluoropentanol-2, 2-pentanol,3,3,4,4,5,5,5-heptafluoro, 2-pentanol, 3,3,4,4,5,5,5-heptafluoro, 3,3,4,4,5,5,5-heptafluoro-2-pentanol, 3,3,4,4,5,5,5-heptakis fluoranyl pentan-2-ol
CC(C(C(C(F)(F)F)(F)F)(F)F)O
214.083
214.08
3,3,4,4,5,5,5-Heptafluoro-2-pentanol
1g
C5H5F7O
1705854
RBPHBIMHZSTIDT-UHFFFAOYSA-N
3,3,4,4,5,5,5-heptafluoropentan-2-ol
78992
96%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning