Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,3,3-Triphenylpropionic acid, 98%

$74.58 - $304.36

Chemical Identifiers

Chemical Name or Material 3,3,3-Triphenylpropionic acid
CAS 900-91-4
Molecular Formula (C6H5)3CCH2CO2H
Formula Weight 302.38
MDL Number MFCD00002713
Beilstein 2057448
Synonym Tritylacetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1668606

 
alfa aesar™
A1668606
5g Each for $74.58

AAA1668614

 
alfa aesar™
A1668614
25g Each for $304.36
Description & Specifications

Specifications

Formula Weight 302.38
Molecular Formula (C6H5)3CCH2CO2H
Quantity 5g
Chemical Name or Material 3,3,3-Triphenylpropionic acid
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 900-91-4
Melting Point 180° to 183°C
MDL Number MFCD00002713
Synonym Tritylacetic acid
Beilstein 2057448

Applications
3,3,3-Triphenylpropionic acid is used as pharmaceutical intermediates.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.