Learn More
(3,3,3-Trifluoropropyl)trimethoxysilane, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1667106
Description
(3,3,3-Trifluoropropyl)trimethoxysilane is used in the manufacture of defoaming agents, releasing agents, fluoro silicone resin and coating. It is used as a coupling agent for epoxy resin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(3,3,3-Trifluoropropyl)trimethoxysilane is used in the manufacture of defoaming agents, releasing agents, fluoro silicone resin and coating. It is used as a coupling agent for epoxy resin.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids, strong bases and strong reducing agents.
Specifications
144°C | |
429-60-7 | |
C6H13F3O3Si | |
UN1993 | |
38°C (100°F) | |
JLGNHOJUQFHYEZ-UHFFFAOYSA-N | |
trimethoxy(3,3,3-trifluoropropyl)silane | |
67933 | |
97% | |
Moisture sensitive |
(3,3,3-Trifluoropropyl)trimethoxysilane | |
5 g | |
MFCD00039266 | |
1762962 | |
trimethoxy 3,3,3-trifluoropropyl silane, 3,3,3-trifluoropropyl trimethoxysilane, 3,3,3-trifluoropropyltrimethoxysilane, silane, trimethoxy 3,3,3-trifluoropropyl, 1,1,1-trifluoro-3-trimethoxysilyl propane, silane, trimethoxy trifluoropropyl, silane,trimethoxy 3,3,3-trifluoropropyl, acmc-1ajbz, fas-3, cf3 ch2 2si och3 3 | |
CO[Si](CCC(F)(F)F)(OC)OC | |
218.25 | |
218.25 | |
1.355 | |
1.14 |
Chemical Identifiers
429-60-7 | |
218.25 | |
JLGNHOJUQFHYEZ-UHFFFAOYSA-N | |
67933 | |
CO[Si](CCC(F)(F)F)(OC)OC |
C6H13F3O3Si | |
MFCD00039266 | |
trimethoxy 3,3,3-trifluoropropyl silane, 3,3,3-trifluoropropyl trimethoxysilane, 3,3,3-trifluoropropyltrimethoxysilane, silane, trimethoxy 3,3,3-trifluoropropyl, 1,1,1-trifluoro-3-trimethoxysilyl propane, silane, trimethoxy trifluoropropyl, silane,trimethoxy 3,3,3-trifluoropropyl, acmc-1ajbz, fas-3, cf3 ch2 2si och3 3 | |
trimethoxy(3,3,3-trifluoropropyl)silane |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 207-059-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only