missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (3,3,3-Trifluoropropyl)trichlorosilane, 97%

$55.20 - $160.38

Chemical Identifiers

CAS 592-09-6
Molecular Formula C3H4Cl3F3Si
Molecular Weight (g/mol) 231.495
MDL Number MFCD00013602
InChI Key WEUBQNJHVBMUMD-UHFFFAOYSA-N
Synonym trichloro 3,3,3-trifluoropropyl silane, 3,3,3-trifluoropropyl trichlorosilane, silane, trichloro 3,3,3-trifluoropropyl, 3,3,3-trifluoropropyltrichlorosilane, acmc-20ajez, cf3ch2ch2sicl3, silane, trichloro trifluoropropyl, 3,3,3-trifluropropyltrichlorosiline, trichloro 3,3,3-trifluoropropyl silane #, trichloro 3,3,3-trifluoropropyl silane
PubChem CID 68963
IUPAC Name trichloro(3,3,3-trifluoropropyl)silane
SMILES C(C[Si](Cl)(Cl)Cl)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1667006 View Documents Alfa Aesar™
L1667006
5g Each for $55.20
 
AAL1667014 View Documents Alfa Aesar™
L1667014
25g Each for $160.38
 
Description

Description

Applications
(3,3,3-Trifluoropropyl)trichlorosilane is used for the production of fluoro silicones liquids.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

592-09-6
231.495
WEUBQNJHVBMUMD-UHFFFAOYSA-N
68963
C(C[Si](Cl)(Cl)Cl)C(F)(F)F
C3H4Cl3F3Si
MFCD00013602
trichloro 3,3,3-trifluoropropyl silane, 3,3,3-trifluoropropyl trichlorosilane, silane, trichloro 3,3,3-trifluoropropyl, 3,3,3-trifluoropropyltrichlorosilane, acmc-20ajez, cf3ch2ch2sicl3, silane, trichloro trifluoropropyl, 3,3,3-trifluropropyltrichlorosiline, trichloro 3,3,3-trifluoropropyl silane #, trichloro 3,3,3-trifluoropropyl silane
trichloro(3,3,3-trifluoropropyl)silane

Specifications

592-09-6
CF3CH2CH2SiCl3
UN2985
trichloro 3,3,3-trifluoropropyl silane, 3,3,3-trifluoropropyl trichlorosilane, silane, trichloro 3,3,3-trifluoropropyl, 3,3,3-trifluoropropyltrichlorosilane, acmc-20ajez, cf3ch2ch2sicl3, silane, trichloro trifluoropropyl, 3,3,3-trifluropropyltrichlorosiline, trichloro 3,3,3-trifluoropropyl silane #, trichloro 3,3,3-trifluoropropyl silane
C(C[Si](Cl)(Cl)Cl)C(F)(F)F
231.495
231.51
113°C to 114°C
15°C (59°F)
C3H4Cl3F3Si
MFCD00013602
1748876
WEUBQNJHVBMUMD-UHFFFAOYSA-N
trichloro(3,3,3-trifluoropropyl)silane
68963
97%
5g
(3,3,3-Trifluoropropyl)trichlorosilane
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CHLOROSILANES, FLAMMABLE, CORROSIVE, N.O.S.

TSCA : TSCA