Please login to your online account to display your discounted pricing

(3,3,3-Trifluoropropyl)chlorodimethylsilane, 95%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC432210250

 View more versions of this product

Catalog No. AC432210250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥94%
Formula Weight 190.67
Quantity 25g
Color Clear colorless
Density 1.11g/mL
CAS Max % 100.0
CAS Min % 94.0
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material (3, 3, 3-Trifluoropropyl)chlorodimethylsilane
Health Hazard 1 GHS Signal Word: Danger
Molecular Formula C5H10ClF3Si
Assay Percent Range 95%
CAS 1481-41-0
Refractive Index 1.3770 to 1.3790 (20°C, 589nm)
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Highly flammable liquid and vapour.
Reacts violently with water.
Contact with water liberates toxic gas.

Specifications

Chemical Name or Material (3, 3, 3-Trifluoropropyl)chlorodimethylsilane
CAS 1481-41-0
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C5H10ClF3Si
Formula Weight 190.67
Accessories

GHS Signal Word: Danger

GHS P Statement:
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Highly flammable liquid and vapour.
Reacts violently with water.
Contact with water liberates toxic gas.