Please login to your online account to display your discounted pricing

(3,3,3-Trifluoropropyl)chlorodimethylsilane, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (3, 3, 3-Trifluoropropyl)chlorodimethylsilane
CAS 1481-41-0
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C5H10ClF3Si
Formula Weight 190.67
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC432210050

 
thermo fisher scientific
AC432210050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC432210250

 
thermo fisher scientific
AC432210250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Formula Weight 190.67
Physical Form Liquid
Quantity 5g
Color Clear colorless
Density 1.11g/mL
CAS Min % 94.0
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Highly flammable liquid and vapour.
Reacts violently with water.
Contact with water liberates toxic gas.
Health Hazard 3 GHS P Statement:
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 95%
CAS 1481-41-0
Chemical Name or Material (3, 3, 3-Trifluoropropyl)chlorodimethylsilane
Refractive Index 1.3770 to 1.3790 (20°C, 589nm)
Molecular Formula C5H10ClF3Si
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Highly flammable liquid and vapour.
Reacts violently with water.
Contact with water liberates toxic gas.