missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(3,3,3-Trifluoropropyl)chlorodimethylsilane, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC432210050

 View more versions of this product

Catalog No. AC432210050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥94%
Liquid
Clear colorless
94.0
95%
(3, 3, 3-Trifluoropropyl)chlorodimethylsilane
C5H10ClF3Si
Authentic
190.67
5g
1.11g/mL
100.0
1481-41-0
1.3770 to 1.3790 (20°C, 589nm)
Glass bottle

Chemical Identifiers

1481-41-0
100.0
C5H10ClF3Si
(3, 3, 3-Trifluoropropyl)chlorodimethylsilane
94.0
95%
190.67
≥94%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
Highly flammable liquid and vapour.
Reacts violently with water.
Contact with water liberates toxic gas.

GHS P Statement:
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger