Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,3,3-Trifluorolactic acid, 96%

Chemical Identifiers

Chemical Name or Material 3,3,3-Trifluorolactic acid
CAS 684-07-1
Molecular Formula C3H3F3O3
Formula Weight 144.05
MDL Number MFCD00051802
UN Number UN3261
Beilstein 1907003
Synonym 3,3,3-Trifluoro-2-hydroxypropionic acid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AAL0774203

  SDS
alfa aesar™
L0774203
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0774206

  SDS
alfa aesar™
L0774206
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 144.05
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Chemical Name or Material 3,3,3-Trifluorolactic acid
CAS 684-07-1
MDL Number MFCD00051802
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 96%
Melting Point 66° to 71°C
Molecular Formula C3H3F3O3
Synonym 3,3,3-Trifluoro-2-hydroxypropionic acid
Quantity 1g
Boiling Point 157°C (123mmHg)
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Beilstein 1907003
UN Number UN3261
Health Hazard 1 Danger

Applications
3,3,3-Trifluorolactic acid is used as pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from oxidizing agents, strong bases and water. Protect from humidity and water. Incompatible with oxidizing agents, bases.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.