missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,3,3-Trifluoro-2-(trifluoromethyl)propionic acid, 97%

Manufacturer:  Alfa Aesar™ H5338803

 View more versions of this product

Catalog No. AAH5338803



Specifications

Specifications

3,3,3-Trifluoro-2-(trifluoromethyl)propionic acid
46°C to 53°C
564-10-3
(CF3)2CHCO2H
3,3,3-trifluoro-2-trifluoromethyl propanoic acid, 2-trifluoromethyl-3,3,3-trifluoropropionic acid, 3,3,3-trifluoro-2-trifluoromethyl propionic acid, propanoic acid,3,3,3-trifluoro-2-trifluoromethyl, hexafluoroisobutyric acid, acmc-1amsz, cf3 2chco2h, 3,3,3-trifluoro-2-trifluoromethylpropionic acid, 3,3,3-trifluoro-2-trifluoromethyl-propionic acid, 3,3,3-tris fluoranyl-2-trifluoromethyl propanoic acid
C(C(=O)O)(C(F)(F)F)C(F)(F)F
196.048
196.05
125°C
1g
C4H2F6O2
MFCD00464165
RAEAYTICAPHWJW-UHFFFAOYSA-N
3,3,3-trifluoro-2-(trifluoromethyl)propanoic acid
385606
97%

Chemical Identifiers

564-10-3
196.048
RAEAYTICAPHWJW-UHFFFAOYSA-N
385606
C(C(=O)O)(C(F)(F)F)C(F)(F)F
C4H2F6O2
MFCD00464165
3,3,3-trifluoro-2-trifluoromethyl propanoic acid, 2-trifluoromethyl-3,3,3-trifluoropropionic acid, 3,3,3-trifluoro-2-trifluoromethyl propionic acid, propanoic acid,3,3,3-trifluoro-2-trifluoromethyl, hexafluoroisobutyric acid, acmc-1amsz, cf3 2chco2h, 3,3,3-trifluoro-2-trifluoromethylpropionic acid, 3,3,3-trifluoro-2-trifluoromethyl-propionic acid, 3,3,3-tris fluoranyl-2-trifluoromethyl propanoic acid
3,3,3-trifluoro-2-(trifluoromethyl)propanoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents