missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3(2H)-Pyridazinone, TCI America™

Manufacturer:  TCI America P185025G

 View more versions of this product

Catalog No. P185025G



Specifications

Specifications

3(2H)-Pyridazinone
White-Yellow
25g
C4H4N2O
3 2h-pyridazinone, pyridazin-3 2h-one, 2h-pyridazin-3-one, 3-hydroxypyridazine, 3-pyridazinol, pyridazin-3-ol, pyridazinone, 2,3-dihydropyridazin-3-one, 3-pyridazinone, 6-pyridazinone
C1=CC(=O)NN=C1
96.089
96.09
≥96.0% (GC,T)
101°C
102°C
504-30-3
MFCD00099605
AAILEWXSEQLMNI-UHFFFAOYSA-N
1H-pyridazin-6-one
68153
Crystal-Powder at 20°C

Chemical Identifiers

504-30-3
96.089
AAILEWXSEQLMNI-UHFFFAOYSA-N
68153
C1=CC(=O)NN=C1
C4H4N2O
MFCD00099605
3 2h-pyridazinone, pyridazin-3 2h-one, 2h-pyridazin-3-one, 3-hydroxypyridazine, 3-pyridazinol, pyridazin-3-ol, pyridazinone, 2,3-dihydropyridazin-3-one, 3-pyridazinone, 6-pyridazinone
1H-pyridazin-6-one
Safety and Handling

Safety and Handling

TSCA : No

Documents