Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(2-Trimethylsiloxyphenyl)-1-propylboronic acid pinacol ester, 95%

Chemical Identifiers

Chemical Name or Material 3-(2-Trimethylsiloxyphenyl)-1-propylboronic acid pinacol ester
Molecular Formula C18H31BO3Si
Formula Weight 334.34
MDL Number MFCD12546185
Synonym 4,4,5,5-Tetramethyl-2-[3-(2-trimethylsiloxyphenyl)propyl]-1,3,2-dioxaborolane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3032303

  SDS
alfa aesar™
H3032303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3032306

  SDS
alfa aesar™
H3032306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3032314

  SDS
alfa aesar™
H3032314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 334.34
Chemical Name or Material 3-(2-Trimethylsiloxyphenyl)-1-propylboronic acid pinacol ester
Quantity 1g
Assay 95%
Molecular Formula C18H31BO3Si
MDL Number MFCD12546185
Synonym 4,4,5,5-Tetramethyl-2-[3-(2-trimethylsiloxyphenyl)propyl]-1,3,2-dioxaborolane