missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-[2-(Trifluoromethyl)phenylsulfamoyl]benzeneboronic acid, 97%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific H54924MD

 View more versions of this product

Catalog No. AAH54924MD



Specifications

Specifications

3-[2-(Trifluoromethyl)phenylsulfamoyl]benzeneboronic acid
1449142-61-3
MFCD20040271
GYJGSTNPUJQMHS-UHFFFAOYSA-N
[3-[[2-(trifluoromethyl)phenyl]sulfamoyl]phenyl]boronic acid
73996034
97%
250mg
C13H11BF3NO4S
3-2-trifluoromethyl phenyl sulfamoyl phenylboronic acid
B(C1=CC(=CC=C1)S(=O)(=O)NC2=CC=CC=C2C(F)(F)F)(O)O
345.099
345.1

Chemical Identifiers

1449142-61-3
345.099
GYJGSTNPUJQMHS-UHFFFAOYSA-N
73996034
B(C1=CC(=CC=C1)S(=O)(=O)NC2=CC=CC=C2C(F)(F)F)(O)O
C13H11BF3NO4S
MFCD20040271
3-2-trifluoromethyl phenyl sulfamoyl phenylboronic acid
[3-[[2-(trifluoromethyl)phenyl]sulfamoyl]phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents