Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-[2-(Trifluoromethyl)phenyl]propionic acid, 97%

$76.54 - $305.88

Chemical Identifiers

Chemical Name or Material 3-[2-(Trifluoromethyl)phenyl]propionic acid
CAS 94022-99-8
Molecular Formula C10H9F3O2
Formula Weight 218.17
MDL Number MFCD06823965
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3187603

  SDS
alfa aesar™
H3187603
1g Each for $76.54

AAH3187606

  SDS
alfa aesar™
H3187606
5g Each for $305.88
Description & Specifications

Specifications

Formula Weight 218.17
Quantity 1g
MDL Number MFCD06823965
Melting Point 79° to 86°C
CAS 94022-99-8
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Assay 97%
Chemical Name or Material 3-[2-(Trifluoromethyl)phenyl]propionic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C10H9F3O2
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.