Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(2-Thienyl)pyridine, 99%

$72.04 - $197.03

Chemical Identifiers

Chemical Name or Material 3-(2-Thienyl)pyridine
CAS 21298-53-3
Molecular Formula C9H7NS
Formula Weight 161.23
MDL Number MFCD01860052
Beilstein 971159
Synonym 2-(3-Pyridyl)thiophene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1853803

 
alfa aesar™
L1853803
1g Each for $197.03

AAL18538MD

 
alfa aesar™
L18538MD
250mg Each for $72.04
Description & Specifications

Specifications

Formula Weight 161.23
Chemical Name or Material 3-(2-Thienyl)pyridine
Assay 99%
MDL Number MFCD01860052
CAS 21298-53-3
Molecular Formula C9H7NS
Beilstein 971159
Synonym 2-(3-Pyridyl)thiophene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Boiling Point 95° to 100°C (1mmHg)
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.