missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(2-Thienyl)-L-alanine, 95%

Manufacturer:  Alfa Aesar™ H63316MD

 View more versions of this product

Catalog No. AAH63316MD


 

Description

Description

Applications
3-(2-Thienyl)-L-alanine can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Specifications

3-(2-Thienyl)-L-alanine
95%
MFCD00064335
H-3-(2-thienyl)-Ala-OH; (S)-2-Amino-3-(2-thienyl)propionic acid
C1=CSC(=C1)CC(C(=O)O)N
171.214
171.21
White
250mg
22951-96-8
C7H9NO2S
82872
WTOFYLAWDLQMBZ-LURJTMIESA-N
(2S)-2-amino-3-thiophen-2-ylpropanoic acid
146719
Solid
255°C to 263°C (decomposition)

Chemical Identifiers

22951-96-8
171.214
WTOFYLAWDLQMBZ-LURJTMIESA-N
146719
C7H9NO2S
MFCD00064335
H-3-(2-thienyl)-Ala-OH; (S)-2-Amino-3-(2-thienyl)propionic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents