missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(2-Thienyl)-D-alanine, 95%

Manufacturer:  Alfa Aesar™ H63681MD

 View more versions of this product

Catalog No. AAH63681MD



Specifications

Specifications

3-(2-Thienyl)-D-alanine
C7H9NO2S
3-2-thienyl-d-alanine, d-2-thienylalanine, r-2-amino-3-thiophen-2-yl propanoic acid, beta-2-thienyl-d-alanine, h-d-thi-oh, 2r-2-amino-3-thiophen-2-ylpropanoic acid, 2r-2-amino-3-thiophen-2-yl propanoic acid, 2r-2-azanyl-3-thiophen-2-yl-propanoic acid, pubchem18712, beta-2-thienyl-d-alanine
C1=CSC(=C1)CC(C(=O)O)N
171.214
CHEBI:77819
Solid
250mg
62561-76-6
MFCD00077166
WTOFYLAWDLQMBZ-ZCFIWIBFSA-N
(2R)-2-amino-3-thiophen-2-ylpropanoic acid
7111750
171.21
95%

Chemical Identifiers

62561-76-6
171.214
WTOFYLAWDLQMBZ-ZCFIWIBFSA-N
7111750
(2R)-2-amino-3-thiophen-2-ylpropanoic acid
C7H9NO2S
MFCD00077166
3-2-thienyl-d-alanine, d-2-thienylalanine, r-2-amino-3-thiophen-2-yl propanoic acid, beta-2-thienyl-d-alanine, h-d-thi-oh, 2r-2-amino-3-thiophen-2-ylpropanoic acid, 2r-2-amino-3-thiophen-2-yl propanoic acid, 2r-2-azanyl-3-thiophen-2-yl-propanoic acid, pubchem18712, beta-2-thienyl-d-alanine
CHEBI:77819
C1=CSC(=C1)CC(C(=O)O)N
Safety and Handling

Safety and Handling

storageNote1 : -30° to -10°C

SDS Documents