Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(2-Thienyl)acrylic acid, predominantly trans, 98+%

$52.59 - $1,020.08

Chemical Identifiers

Chemical Name or Material 3- (2-Thienyl)acrylic acid
CAS 15690-25-2
Name Note predominantly trans
Molecular Formula C7H6O2S
Formula Weight 154.19
MDL Number MFCD00005455
Beilstein 112053
Synonym Thiophene-2-acrylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1034209

  SDS
alfa aesar™
A1034209
10g Each for $52.59

AAA1034218

  SDS
alfa aesar™
A1034218
50g Each for $241.08

AAA1034230

  SDS
alfa aesar™
A1034230
250g Each for $1,020.08
Description & Specifications

Specifications

Formula Weight 154.19
Health Hazard 1 Warning
CAS 15690-25-2
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 145° to 148°C
Molecular Formula C7H6O2S
Chemical Name or Material 3- (2-Thienyl)acrylic acid
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00005455
Name Note predominantly trans
Assay ≥98%
Quantity 10g
Beilstein 112053
Synonym Thiophene-2-acrylic acid
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.