missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(2-Thienyl)-1-(p-tolyl)-2-propen-1-one, 98%

Manufacturer:  Alfa Aesar™ A1108314

 View more versions of this product

Catalog No. AAA1108314


 

Specifications

Specifications

3- (2-Thienyl)-1- (p-tolyl)-2-propen-1-one
25g
98%
MFCD00022532
OUDDTWPBSXBJOY-CMDGGOBGSA-N
(E)-1-(4-methylphenyl)-3-thiophen-2-ylprop-2-en-1-one
642339
78°C to 80°C
6028-89-3
C14H12OS
1-(4-Methylphenyl)-3-(2-thienyl)-prop-2-en-1-one
CC1=CC=C(C=C1)C(=O)C=CC2=CC=CS2
228.309
228.32

Chemical Identifiers

6028-89-3
228.309
OUDDTWPBSXBJOY-CMDGGOBGSA-N
642339
C14H12OS
MFCD00022532
1-(4-Methylphenyl)-3-(2-thienyl)-prop-2-en-1-one
SDS Documents