Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(2-Pyrrolidinyl)piperidine, 94%

Chemical Identifiers

Chemical Name or Material 3-(2-Pyrrolidinyl)piperidine
CAS 858262-19-8
Molecular Formula C9H18N2
Formula Weight 154.26
MDL Number MFCD08277606
UN Number UN3259
Synonym 2-(3-Piperidinyl)pyrrolidine; 3-(2-Pyrrolidino)piperidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH26049MC

  SDS
alfa aesar™
H26049MC
100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 154.26
Chemical Name or Material 3-(2-Pyrrolidinyl)piperidine
Assay 94%
Molecular Formula C9H18N2
MDL Number MFCD08277606
CAS 858262-19-8
UN Number UN3259
Synonym 2-(3-Piperidinyl)pyrrolidine; 3-(2-Pyrrolidino)piperidine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
Quantity 100mg
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.