missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(2-Pyridyl)-L-alanine dihydrochloride, 95%

$48.58 - $573.66

Chemical Identifiers

CAS 37535-51-6
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD06809729
InChI Key PDRJLZDUOULRHE-ZETCQYMHSA-N
Synonym l-2-pyridylalanine, 3-2-pyridyl-l-alanine, s-2-amino-3-pyridin-2-yl propanoic acid, h-2-pal-oh, l-3-2-pyridyl-alanine, 3-2-pyridyl-alanine, 2'-pyridyl-l-ala, 2s-2-amino-3-pyridin-2-yl propanoic acid, 2-aza-d-phenylalanine, 3-pyridin-2-yl-d-alanine
PubChem CID 2761499
IUPAC Name (2S)-2-amino-3-pyridin-2-ylpropanoic acid
SMILES C1=CC=NC(=C1)CC(C(=O)O)N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63770MD View Documents Alfa Aesar™
H63770MD
250mg Each for $48.58
 
AAH6377003 View Documents Alfa Aesar™
H6377003
1g Each for $144.04
 
AAH6377006 View Documents Alfa Aesar™
H6377006
5g Each for $573.66
 
Specifications

Chemical Identifiers

37535-51-6
166.18
PDRJLZDUOULRHE-ZETCQYMHSA-N
2761499
C1=CC=NC(=C1)CC(C(=O)O)N
C8H10N2O2
MFCD06809729
l-2-pyridylalanine, 3-2-pyridyl-l-alanine, s-2-amino-3-pyridin-2-yl propanoic acid, h-2-pal-oh, l-3-2-pyridyl-alanine, 3-2-pyridyl-alanine, 2'-pyridyl-l-ala, 2s-2-amino-3-pyridin-2-yl propanoic acid, 2-aza-d-phenylalanine, 3-pyridin-2-yl-d-alanine
(2S)-2-amino-3-pyridin-2-ylpropanoic acid

Specifications

37535-51-6
MFCD06809729
PDRJLZDUOULRHE-ZETCQYMHSA-N
(2S)-2-amino-3-pyridin-2-ylpropanoic acid
2761499
Solid
250mg
C8H10N2O2
l-2-pyridylalanine, 3-2-pyridyl-l-alanine, s-2-amino-3-pyridin-2-yl propanoic acid, h-2-pal-oh, l-3-2-pyridyl-alanine, 3-2-pyridyl-alanine, 2'-pyridyl-l-ala, 2s-2-amino-3-pyridin-2-yl propanoic acid, 2-aza-d-phenylalanine, 3-pyridin-2-yl-d-alanine
C1=CC=NC(=C1)CC(C(=O)O)N
166.18
239.1
95%
3-(2-Pyridyl)-L-alanine dihydrochloride