missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-(2-Pyridyl)-D-alanine, 95%, Thermo Scientific™
$281.23 - $281.23
Chemical Identifiers
CAS | 37535-52-7 |
---|---|
Molecular Formula | C8H10N2O2 |
Molecular Weight (g/mol) | 166.18 |
MDL Number | MFCD00237245 |
InChI Key | PDRJLZDUOULRHE-SSDOTTSWSA-N |
Synonym | 3-2-pyridyl-d-alanine, r-2-amino-3-pyridin-2-yl propanoic acid, d-2-pyridylalanine, 2r-2-amino-3-pyridin-2-yl propanoic acid, h-d-2-pal-oh, 2'-pyridyl-d-ala, 3-2-pyridyl-d-ala-oh, d-3-2-pyridyl-alanine, r-2-pyridylalanine, 2-pyridyl-d-ala |
PubChem CID | 7019768 |
IUPAC Name | (2R)-2-amino-3-pyridin-2-ylpropanoic acid |
SMILES | C1=CC=NC(=C1)CC(C(=O)O)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6329103
|
Thermo Scientific Chemicals
H6329103 |
1 g |
Each for $281.23
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
37535-52-7 | |
166.18 | |
PDRJLZDUOULRHE-SSDOTTSWSA-N | |
7019768 | |
C1=CC=NC(=C1)CC(C(=O)O)N |
C8H10N2O2 | |
MFCD00237245 | |
3-2-pyridyl-d-alanine, r-2-amino-3-pyridin-2-yl propanoic acid, d-2-pyridylalanine, 2r-2-amino-3-pyridin-2-yl propanoic acid, h-d-2-pal-oh, 2'-pyridyl-d-ala, 3-2-pyridyl-d-ala-oh, d-3-2-pyridyl-alanine, r-2-pyridylalanine, 2-pyridyl-d-ala | |
(2R)-2-amino-3-pyridin-2-ylpropanoic acid |
Specifications
37535-52-7 | |
MFCD00237245 | |
3-2-pyridyl-d-alanine, r-2-amino-3-pyridin-2-yl propanoic acid, d-2-pyridylalanine, 2r-2-amino-3-pyridin-2-yl propanoic acid, h-d-2-pal-oh, 2'-pyridyl-d-ala, 3-2-pyridyl-d-ala-oh, d-3-2-pyridyl-alanine, r-2-pyridylalanine, 2-pyridyl-d-ala | |
C1=CC=NC(=C1)CC(C(=O)O)N | |
166.18 | |
166.18 | |
1 g | |
3-(2-Pyridyl)-D-alanine |
C8H10N2O2 | |
6478361 | |
PDRJLZDUOULRHE-SSDOTTSWSA-N | |
(2R)-2-amino-3-pyridin-2-ylpropanoic acid | |
7019768 | |
95% | |
Solid |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures