missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(2-Pyridyl)-D-alanine, 95%

Manufacturer:  Alfa Aesar™ H6329106

 View more versions of this product

Catalog No. AAH6329106



Specifications

Specifications

3-(2-Pyridyl)-D-alanine
C8H10N2O2
6478361
PDRJLZDUOULRHE-SSDOTTSWSA-N
(2R)-2-amino-3-pyridin-2-ylpropanoic acid
7019768
Solid
5g
37535-52-7
MFCD00237245
3-2-pyridyl-d-alanine, r-2-amino-3-pyridin-2-yl propanoic acid, d-2-pyridylalanine, 2r-2-amino-3-pyridin-2-yl propanoic acid, h-d-2-pal-oh, 2'-pyridyl-d-ala, 3-2-pyridyl-d-ala-oh, d-3-2-pyridyl-alanine, r-2-pyridylalanine, 2-pyridyl-d-ala
C1=CC=NC(=C1)CC(C(=O)O)N
166.18
166.18
95%

Chemical Identifiers

37535-52-7
166.18
PDRJLZDUOULRHE-SSDOTTSWSA-N
7019768
C1=CC=NC(=C1)CC(C(=O)O)N
C8H10N2O2
MFCD00237245
3-2-pyridyl-d-alanine, r-2-amino-3-pyridin-2-yl propanoic acid, d-2-pyridylalanine, 2r-2-amino-3-pyridin-2-yl propanoic acid, h-d-2-pal-oh, 2'-pyridyl-d-ala, 3-2-pyridyl-d-ala-oh, d-3-2-pyridyl-alanine, r-2-pyridylalanine, 2-pyridyl-d-ala
(2R)-2-amino-3-pyridin-2-ylpropanoic acid
Documents