Please login to your online account to display your discounted pricing

3-[2-Oxo-3-(trifluoromethyl)-1,2-dihydropyridin-1-yl]propanenitrile, 97%, Maybridge


Quantity 1g

Manufacturer: fisher scientific company  SPB06060DA

 View more versions of this product

Catalog No. SPB06060DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 216.16
Quantity 1g
Assay 97%
CAS Min % 97.0
Chemical Name or Material 3-[2-Oxo-3-(trifluoromethyl)-1, 2-dihydropyridin-1-yl]propanenitrile
Assay Percent Range 97%
Molecular Formula C9H7F3N2O
MDL Number MFCD00204216
CAS 175277-60-8
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.

Specifications

Chemical Name or Material 3-[2-Oxo-3-(trifluoromethyl)-1, 2-dihydropyridin-1-yl]propanenitrile
CAS 175277-60-8
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H7F3N2O
Formula Weight 216.16
MDL Number MFCD00204216
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.

SDS & Safety Documents