Please login to your online account to display your discounted pricing

3-(2-Oxo-2,3-dihydro-1,3-benzoxazol-3-yl)propanoic acid, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  SPB02696EA

 View more versions of this product

Catalog No. SPB02696EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 207.18
Quantity 10g
Assay 97%
CAS Min % 97.0
Molecular Formula C10H9NO4
CAS 13610-59-8
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 3-(2-Oxo-2, 3-dihydro-1, 3-benzoxazol-3-yl)propanoic acid
Assay Percent Range 97%
MDL Number MFCD00052166
Packaging Amber Glass Bottle
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Specifications

Chemical Name or Material 3-(2-Oxo-2, 3-dihydro-1, 3-benzoxazol-3-yl)propanoic acid
CAS 13610-59-8
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H9NO4
Formula Weight 207.18
MDL Number MFCD00052166
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.