Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(2-Naphthylthio)propionic acid, 97%

$95.28 - $320.18

Chemical Identifiers

Chemical Name or Material 3-(2-Naphthylthio)propionic acid
CAS 1141-45-3
Molecular Formula C13H12O2S
Formula Weight 232.3
MDL Number MFCD00051616
Beilstein 2558498
Synonym 3-(2-Naphthylmercapto)propionic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0620803

 
alfa aesar™
L0620803
1g Each for $95.28

AAL0620806

 
alfa aesar™
L0620806
5g Each for $320.18
Description & Specifications

Specifications

Formula Weight 232.3
Chemical Name or Material 3-(2-Naphthylthio)propionic acid
Molecular Formula C13H12O2S
Assay 97%
Melting Point 103° to 107°C
MDL Number MFCD00051616
CAS 1141-45-3
Beilstein 2558498
Synonym 3-(2-Naphthylmercapto)propionic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.