Please login to your online account to display your discounted pricing

3-(2-Naphthyl)-L-alanine, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  333900010

 View more versions of this product

Catalog No. AC333900010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 215.25
Physical Form Crystalline Powder
Quantity 1g
Color Off-white
Assay Percent Range 98%
CAS 58438-03-2
Molecular Formula C13H13NO2
Chemical Name or Material 3-(2-Naphthyl)-L-alanine
MDL Number MFCD00066087
CAS Max % 100.0
CAS Min % 97.5
Packaging Glass bottle
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 3-(2-Naphthyl)-L-alanine
CAS 58438-03-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C13H13NO2
Formula Weight 215.25
MDL Number MFCD00066087